Assay Details
Target Gene Details
Entrez Gene ID: | 55968 |
Gene Name: | NSFL1 cofactor |
Gene Aliases: |
P47, UBX1, UBXD10, UBXN2C, dJ776F14.1 |
Location: |
Chr.20:1442162-1467720 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NSFL1C | NM_001206736.1 | 10 | 3102 | NP_001193665.1 |
| NM_016143.4 | 9 | 3096 | NP_057227.2 | |
| NM_018839.4 | 8 | 3003 | NP_061327.2 | |
| NR_038164.1 | 8 | 2903 | ||
| XM_006723592.2 | 10 | 3035 | XP_006723655.1 | |
| XM_006723593.2 | 9 | 2937 | XP_006723656.1 | |
| XM_006723594.2 | 9 | 3029 | XP_006723657.1 | |
| XM_011529300.1 | 11 | 3139 | XP_011527602.1 | |
| XM_011529301.1 | 10 | 3133 | XP_011527603.1 | |
| XM_011529302.2 | 9 | 3653 | XP_011527604.1 | |
| XM_011529304.1 | 10 | 2340 | XP_011527606.1 | |
| XM_017027956.1 | 8 | 3643 | XP_016883445.1 | |
| XM_017027957.1 | 11 | 3237 | XP_016883446.1 | |
| XM_017027958.1 | 7 | 2166 | XP_016883447.1 | |
| AK001511.1 | 8 | 2139 | BAA91731.1 | |
| AK128722.1 | 2 | 3342 | BAC87590.1 | |
| BQ007132.1 | 1 | 483 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522008 | Chr.20:1359746 - 1657932 on Build GRCh38 | Gain |
|
| nsv833893 | Chr.20:1433971 - 1547133 on Build GRCh38 | Loss |
|
| nsv585206 | Chr.20:1405829 - 1733716 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map