Assay Details
Target Gene Details
Entrez Gene ID: | 26256 |
Gene Name: | calcium binding tyrosine phosphorylation regulated |
Gene Aliases: |
CABYRa, CABYRc, CABYRc/d, CABYRe, CBP86, CT88, FSP-2, FSP2 |
Location: |
Chr.18:24138956-24161600 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABYR | NM_001308231.1 | NP_001295160.1 | ||
| NM_012189.3 | NP_036321.2 | |||
| NM_138643.2 | NP_619584.1 | |||
| NM_138644.2 | NP_619585.1 | |||
| NM_153768.2 | NP_722452.1 | |||
| NM_153769.2 | NP_722453.1 | |||
| NM_153770.2 | NP_722454.1 | |||
| XM_005258247.2 | XP_005258304.1 | |||
| XM_017025706.1 | XP_016881195.1 | |||
| AF088868.1 | AAC35373.1 | |||
| AF295037.1 | AAG17889.1 | |||
| AF295038.1 | AAG17890.1 | |||
| AF295039.1 | AAG17891.1 | |||
| AF329634.1 | AAL56051.1 | |||
| AK225858.1 | ||||
| AK301889.1 | ||||
| AK313243.1 | ||||
| BC011996.1 | AAH11996.1 | |||
| DB055487.1 | ||||
| HM594164.1 | ||||
| HM594165.1 | ||||
| HM594166.1 | ||||
| HM594168.1 | ||||
| HQ659768.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76208354] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map