Assay Details
Target Gene Details
Entrez Gene ID: | 5715 |
Gene Name: | proteasome 26S subunit, non-ATPase 9 |
Gene Aliases: |
Rpn4, p27 |
Location: |
Chr.12:121888731-121917865 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD9 | NM_001261400.2 | 4 | 1762 | NP_001248329.1 |
| NM_002813.6 | 6 | 2077 | NP_002804.2 | |
| NR_048555.2 | 5 | 1865 | ||
| BC002383.2 | 6 | 2037 | AAH02383.1 | |
| BC004184.2 | 6 | 2037 | AAH04184.1 | |
| BC004213.2 | 6 | 2037 | AAH04213.1 | |
| BU726372.1 | 1 | 292 |
More Information
Set Membership: |
|

Genomic Map