Assay Details
Target Gene Details
Entrez Gene ID: | 51765 |
Gene Name: | serine/threonine protein kinase 26 |
Gene Aliases: |
MASK, MST4 |
Location: |
Chr.X:132023217-132075943 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK26 | NM_001042453.1 | 1 | 112 | NP_001035918.1 |
| NM_016542.3 | 1 | 112 | NP_057626.2 | |
| XM_011531349.1 | XP_011529651.1 | |||
| XM_011531350.1 | XP_011529652.1 | |||
| AB040057.2 | BAA92785.2 | |||
| AF231012.1 | AAK38484.1 | |||
| AK075107.1 | BAC11406.1 | |||
| AK075150.1 | BAC11435.1 | |||
| AK303583.1 | ||||
| AK314356.1 | ||||
| AK316137.1 | ||||
| BC098315.1 | AAH98315.1 | |||
| BC099843.1 | AAH99843.1 | |||
| BC103503.1 | AAI03504.1 | |||
| DA760264.1 | 1 | 115 |
More Information
Set Membership: |
|

Genomic Map