Assay Details
Target Gene Details
Entrez Gene ID: | 472 |
Gene Name: | ATM serine/threonine kinase |
Gene Aliases: |
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1 |
Location: |
Chr.11:108222484-108369102 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATM | XM_011542843.2 | 1 | 117 | XP_011541145.1 |
| XM_017017791.1 | 1 | 117 | XP_016873280.1 | |
| XM_017017792.1 | 1 | 117 | XP_016873281.1 | |
| X91196.1 | 1 | 117 | CAA62603.1 |
Target Gene Details
Entrez Gene ID: | 4863 |
Gene Name: | nuclear protein, coactivator of histone transcription |
Gene Aliases: |
E14, E14/NPAT, p220 |
Location: |
Chr.11:108155280-108222638 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NPAT | NM_001321307.1 | NP_001308236.1 | ||
| NM_002519.2 | NP_002510.2 | |||
| XM_011542854.2 | XP_011541156.1 | |||
| AK290020.1 | ||||
| AK291687.1 | ||||
| BC040356.1 | AAH40356.1 | |||
| BC050561.2 | AAH50561.1 | |||
| BC136270.1 | ||||
| D83243.1 | BAA11861.1 | |||
| DA338219.1 | ||||
| X97186.1 | CAA65824.1 |
More Information
Set Membership: |
|

Genomic Map