Assay Details
Target Gene Details
Entrez Gene ID: | 92346 |
Gene Name: | chromosome 1 open reading frame 105 |
Gene Aliases: |
- |
Location: |
Chr.1:172418560-172468829 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf105 | NM_139240.3 | NP_640333.3 | ||
| XM_011510152.2 | XP_011508454.1 | |||
| XM_011510153.2 | XP_011508455.1 | |||
| XM_011510154.2 | XP_011508456.1 | |||
| XM_011510155.2 | XP_011508457.1 | |||
| XM_011510156.2 | XP_011508458.1 | |||
| AL035295.1 | CAA22895.1 | |||
| BC038410.1 | AAH38410.1 |
Target Gene Details
Entrez Gene ID: | 5279 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class C |
Gene Aliases: |
GPI2 |
Location: |
Chr.1:172441457-172444090 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGC | NM_002642.3 | 2 | 1048 | NP_002633.1 |
| NM_153747.1 | 2 | 1025 | NP_714969.1 | |
| AK308201.1 | 2 | 1004 | ||
| BC006539.2 | 2 | 995 | AAH06539.1 | |
| BT006734.1 | 1 | 714 | AAP35380.1 | |
| CR450292.1 | 1 | 714 | CAG29288.1 | |
| D85418.1 | 1 | 1007 | BAA12812.1 |
More Information
Set Membership: |
|

Genomic Map