Assay Details
Target Gene Details
Entrez Gene ID: | 347169 |
Gene Name: | olfactory receptor family 1 subfamily B member 1 (gene/pseudogene) |
Gene Aliases: |
OR9-26, OR9-B |
Location: |
Chr.9:122578405-122657626 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR1B1 | NM_001004450.1 | 1 | 169 | NP_001004450.1 |
| XM_017014695.1 | 2 | 698 | XP_016870184.1 | |
| XM_017014696.1 | 2 | 730 | XP_016870185.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1054800 | Chr.9:122555206 - 122666496 on Build GRCh38 | Loss |
|
| nsv1039785 | Chr.9:122544123 - 122644710 on Build GRCh38 | Gain |
|
| nsv6705 | Chr.9:122617748 - 122648067 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map