Assay Details
Target Gene Details
Entrez Gene ID: | 3187 |
Gene Name: | heterogeneous nuclear ribonucleoprotein H1 (H) |
Gene Aliases: |
HNRPH, HNRPH1, hnRNPH |
Location: |
Chr.5:179614178-179634784 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv823357 | Chr.5:179619317 - 179631192 on Build GRCh38 | Gain |
|
| dgv214n111 | Chr.5:179620185 - 179696634 on Build GRCh38 | Duplication |
|
| nsv830560 | Chr.5:179612211 - 179748757 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map