Assay Details
Target Gene Details
Entrez Gene ID: | 55184 |
Gene Name: | double zinc ribbon and ankyrin repeat domains 1 |
Gene Aliases: |
ANKRD64, C20orf12, C20orf84, bA189K21.1, bA189K21.8, dJ568F9.2 |
Location: |
Chr.20:18383367-18467185 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645437 | Chr.20:18357770 - 18484726 on Build GRCh38 | Gain |
|
| dgv4283n100 | Chr.20:18364565 - 18647377 on Build GRCh38 | Loss |
|
| nsv1062244 | Chr.20:18184227 - 18520274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map