Assay Details
Target Gene Details
Entrez Gene ID: | 290 |
Gene Name: | alanyl aminopeptidase, membrane |
Gene Aliases: |
APN, CD13, GP150, LAP1, P150, PEPN |
Location: |
Chr.15:89784895-89814852 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANPEP | NM_001150.2 | 21 | 3352 | NP_001141.2 |
| XM_005254892.4 | 21 | 3360 | XP_005254949.1 | |
| XM_011521473.1 | 21 | 3503 | XP_011519775.1 | |
| AB209918.1 | 21 | 3348 | BAD93155.1 | |
| AK298134.1 | ||||
| AK298339.1 | 16 | 1933 | ||
| AK300946.1 | 17 | 2083 | ||
| BC058928.1 | 20 | 3153 | AAH58928.1 | |
| M22324.1 | 20 | 3180 | AAA51719.1 | |
| X13276.1 | 20 | 3084 | CAA31640.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv819621 | Chr.15:89784895 - 89785200 on Build GRCh38 | Gain |
|
| nsv570388 | Chr.15:89730911 - 89794962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map