Assay Details
Target Gene Details
Entrez Gene ID: | 158427 |
Gene Name: | thiosulfate sulfurtransferase like domain containing 2 |
Gene Aliases: |
C9orf97 |
Location: |
Chr.9:97600080-97633680 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSTD2 | NM_139246.4 | NP_640339.4 | ||
| XM_011518312.2 | XP_011516614.1 | |||
| XM_017014342.1 | XP_016869831.1 | |||
| AF258575.1 | AAG23778.1 | |||
| AK056453.1 | BAB71188.1 | |||
| AK056752.1 | BAB71271.1 | |||
| AK094043.1 | ||||
| AK122809.1 | ||||
| AK128615.1 | BAC87530.1 | |||
| AK291199.1 | ||||
| BC035604.1 | AAH35604.1 | |||
| BX538308.1 | ||||
| BX648493.1 | ||||
| KU178877.1 | ||||
| KU178878.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1494e214 | Chr.9:97595020 - 97658603 on Build GRCh38 | Gain |
|
| esv3621190 | Chr.9:97570964 - 97628055 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map