Assay Details
Target Gene Details
Entrez Gene ID: | 84101 |
Gene Name: | ubiquitin specific peptidase 44 |
Gene Aliases: |
- |
Location: |
Chr.12:95516560-95551529 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP44 | NM_001042403.2 | NP_001035862.1 | ||
| NM_001278393.1 | NP_001265322.1 | |||
| NM_032147.4 | NP_115523.2 | |||
| NR_125360.1 | ||||
| XM_005269172.1 | XP_005269229.1 | |||
| XM_005269173.1 | XP_005269230.1 | |||
| XM_005269174.1 | XP_005269231.1 | |||
| XM_011538800.2 | XP_011537102.1 | |||
| XM_011538801.1 | XP_011537103.1 | |||
| XM_011538802.1 | XP_011537104.1 | |||
| XM_011538803.1 | XP_011537105.1 | |||
| XM_011538804.2 | XP_011537106.1 | |||
| XM_011538805.1 | XP_011537107.1 | |||
| XM_017020009.1 | XP_016875498.1 | |||
| XM_017020010.1 | XP_016875499.1 | |||
| XM_017020011.1 | XP_016875500.1 | |||
| AK027434.1 | ||||
| AK315697.1 | ||||
| AL136825.1 | CAB66759.1 | |||
| BC030704.1 | AAH30704.1 | |||
| HY021046.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580379 | Chr.12:95518596 - 95528911 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75908742] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map