Assay Details
Target Gene Details
Entrez Gene ID: | 57084 |
Gene Name: | solute carrier family 17 member 6 |
Gene Aliases: |
DNPI, VGLUT2 |
Location: |
Chr.11:22338121-22379503 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A6 | NM_020346.2 | NP_065079.1 | ||
| AB032435.1 | BAA92874.1 | |||
| DA392769.1 | 1 | 67 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038630 | Chr.11:22024274 - 22450043 on Build GRCh38 | Loss |
|
| nsv1041233 | Chr.11:22034459 - 22388429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map