Assay Details
Target Gene Details
Entrez Gene ID: | 400823 |
Gene Name: | family with sequence similarity 177 member B |
Gene Aliases: |
- |
Location: |
Chr.1:222737216-222772193 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM177B | NM_001324080.1 | 5 | 689 | NP_001311009.1 |
| NM_207468.2 | 6 | 791 | NP_997351.2 | |
| NR_136691.1 | 6 | 1295 | ||
| XM_017001279.1 | 5 | 1233 | XP_016856768.1 | |
| XM_017001280.1 | 6 | 747 | XP_016856769.1 | |
| XM_017001281.1 | 6 | 1316 | XP_016856770.1 | |
| XM_017001282.1 | 6 | 772 | XP_016856771.1 | |
| XM_017001285.1 | XP_016856774.1 | |||
| AK125494.1 | 6 | 1295 | BAC86182.1 |

Genomic Map