Assay Details
Target Gene Details
Entrez Gene ID: | 92335 |
Gene Name: | STE20-related kinase adaptor alpha |
Gene Aliases: |
LYK5, NY-BR-96, PMSE, STRAD, Stlk |
Location: |
Chr.17:63702832-63741970 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRADA | NM_001003786.2 | NP_001003786.1 | ||
| NM_001003787.2 | NP_001003787.1 | |||
| NM_001003788.2 | NP_001003788.1 | |||
| NM_001165969.1 | NP_001159441.1 | |||
| NM_001165970.1 | NP_001159442.1 | |||
| NM_153335.5 | 9 | 1552 | NP_699166.2 | |
| XM_005257797.2 | XP_005257854.1 | |||
| XM_005257798.2 | XP_005257855.1 | |||
| XM_005257799.2 | XP_005257856.1 | |||
| XM_005257800.2 | XP_005257857.1 | |||
| XM_011525466.2 | XP_011523768.1 | |||
| XM_011525467.2 | XP_011523769.1 | |||
| XM_017025312.1 | XP_016880801.1 | |||
| XM_017025313.1 | XP_016880802.1 | |||
| AF308302.1 | AAG48269.1 | |||
| AK074771.1 | BAC11197.1 | |||
| AK075005.1 | 9 | 1381 | BAC11349.1 | |
| AK128173.1 | 1 | 2473 | ||
| AK293160.1 | ||||
| AK301331.1 | ||||
| AL832407.1 | CAI46194.1 | |||
| AY290821.2 | 9 | 1552 | AAP42280.1 | |
| BC043641.1 | 10 | 1742 | AAH43641.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv575854 | Chr.17:63702567 - 63708544 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map