Assay Details
Target Gene Details
Entrez Gene ID: | 126393 |
Gene Name: | heat shock protein family B (small) member 6 |
Gene Aliases: |
HEL55, Hsp20, PPP1R91 |
Location: |
Chr.19:35754566-35757029 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB6 | NM_144617.2 | 2 | 291 | NP_653218.1 |
| AK056951.1 | 2 | 291 | BAB71323.1 | |
| AK297852.1 | 2 | 286 | ||
| AK300695.1 | ||||
| BC068046.1 | 2 | 279 | AAH68046.1 | |
| EU794622.1 | 2 | 291 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953286 | Chr.19:35747000 - 35868398 on Build GRCh38 | Deletion |
|
| nsv579422 | Chr.19:35665686 - 35774606 on Build GRCh38 | Loss |
|
| nsv579423 | Chr.19:35730150 - 35792945 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map