Assay Details
Target Gene Details
Entrez Gene ID: | 55388 |
Gene Name: | minichromosome maintenance 10 replication initiation factor |
Gene Aliases: |
CNA43, DNA43, PRO2249 |
Location: |
Chr.10:13161554-13211110 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM10 | NM_018518.4 | 5 | 631 | NP_060988.3 |
| NM_182751.2 | 5 | 634 | NP_877428.1 | |
| XM_011519538.2 | 5 | 604 | XP_011517840.1 | |
| AB042719.1 | 5 | 598 | BAB18723.1 | |
| AK055695.1 | 5 | 607 | BAB70988.1 | |
| AK292701.1 | 5 | 609 | ||
| AL136840.1 | 4 | 597 | CAB66774.2 | |
| BC101727.1 | 5 | 603 | AAI01728.1 | |
| BC143490.1 | 5 | 596 |
More Information
Set Membership: |
|

Genomic Map