Assay Details
Target Gene Details
Entrez Gene ID: | 4055 |
Gene Name: | lymphotoxin beta receptor |
Gene Aliases: |
CD18, D12S370, LT-BETA-R, TNF-R-III, TNFCR, TNFR-RP, TNFR2-RP, TNFR3, TNFRSF3 |
Location: |
Chr.12:6375368-6391571 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LTBR | NM_001270987.1 | 10 | 1373 | NP_001257916.1 |
| NM_002342.2 | 10 | 1568 | NP_002333.1 | |
| XM_005253688.1 | 9 | 1420 | XP_005253745.1 | |
| AK027080.1 | 10 | 1433 | ||
| AK095418.1 | 11 | 1397 | ||
| AK128019.1 | 2 | 1627 | ||
| AK293187.1 | 10 | 1312 | ||
| BC026262.1 | 10 | 1440 | AAH26262.1 | |
| BX648074.1 | 2 | 1486 | ||
| L04270.1 | 10 | 1410 | AAA36757.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952790 | Chr.12:6368835 - 6394034 on Build GRCh38 | Deletion |
|
| esv34325 | Chr.12:6342424 - 6513183 on Build GRCh38 | Gain |
|
| esv2759876 | Chr.12:6344718 - 6554669 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map