Assay Details
Target Gene Details
Entrez Gene ID: | 4259 |
Gene Name: | microsomal glutathione S-transferase 3 |
Gene Aliases: |
GST-III |
Location: |
Chr.1:165630873-165656136 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MGST3 | NM_004528.3 | NP_004519.1 | ||
| XM_005245174.3 | XP_005245231.2 | |||
| AF026977.1 | AAB82609.1 | |||
| AK223012.1 | BAD96732.1 | |||
| AK312103.1 | ||||
| BC000505.2 | AAH00505.1 | |||
| BC003034.2 | AAH03034.1 | |||
| BC005964.1 | AAH05964.1 | |||
| BI666647.1 | ||||
| CR450359.1 | CAG29355.1 |
More Information
Set Membership: |
|

Genomic Map