Assay Details
Target Gene Details
Entrez Gene ID: | 6534 |
Gene Name: | solute carrier family 6 member 7 |
Gene Aliases: |
PROT |
Location: |
Chr.5:150189861-150211072 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A7 | NM_014228.4 | 5 | 1126 | NP_055043.2 |
| XM_017009767.1 | 6 | 1033 | XP_016865256.1 | |
| XM_017009768.1 | 6 | 1109 | XP_016865257.1 | |
| XM_017009769.1 | 6 | 2640 | XP_016865258.1 | |
| XM_017009770.1 | 6 | 760 | XP_016865259.1 | |
| AK096607.1 | 5 | 1030 | ||
| AK308895.1 | 5 | 902 | ||
| AK312309.1 | 5 | 906 | ||
| BC069631.1 | 5 | 659 | AAH69631.1 | |
| BC093785.1 | 5 | 735 | AAH93785.1 | |
| BC113425.1 | 5 | 735 | ||
| S80071.1 | 5 | 659 | AAB47007.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762556 | Chr.5:150141049 - 150209450 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map