Assay Details
Target Gene Details
Entrez Gene ID: | 388695 |
Gene Name: | LysM domain containing 1 |
Gene Aliases: |
SB145 |
Location: |
Chr.1:151148496-151165948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYSMD1 | NM_001136543.1 | NP_001130015.1 | ||
| NM_212551.4 | 1 | 370 | NP_997716.1 | |
| AK298706.1 | ||||
| BC031649.1 | AAH31649.1 | |||
| BX647911.1 | 1 | 337 | CAH10574.1 | |
| DA176882.1 | 1 | 373 |
Target Gene Details
Entrez Gene ID: | 100534012 |
Gene Name: | TNFAIP8L2-SCNM1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:151156629-151170297 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFAIP8L2-SCNM1 | NM_001204848.1 | NP_001191777.1 | ||
| BG531092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map