Assay Details
Target Gene Details
Entrez Gene ID: | 283111 |
Gene Name: | olfactory receptor family 51 subfamily V member 1 |
Gene Aliases: |
OR11-36, OR51A12 |
Location: |
Chr.11:5199735-5200700 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR51V1 | NM_001004760.2 | 1 | 906 | NP_001004760.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1048350 | Chr.11:5168066 - 5204612 on Build GRCh38 | Gain |
|
| esv2760169 | Chr.11:4940288 - 5212591 on Build GRCh38 | Gain+Loss |
|
| esv3891916 | Chr.11:5169244 - 5227774 on Build GRCh38 | Loss |
|
| dgv182e214 | Chr.11:5193527 - 5224501 on Build GRCh38 | Loss |
|
| esv3891917 | Chr.11:5081246 - 5227774 on Build GRCh38 | Loss |
|
| nsv523914 | Chr.11:5186159 - 5210335 on Build GRCh38 | Loss |
|
| dgv1041n100 | Chr.11:5154247 - 5248100 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map