Assay Details
Target Gene Details
Entrez Gene ID: | 83854 |
Gene Name: | angiopoietin like 6 |
Gene Aliases: |
AGF, ARP5 |
Location: |
Chr.19:10092337-10106194 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANGPTL6 | NM_001321411.1 | 6 | 1657 | NP_001308340.1 |
| NM_031917.2 | 6 | 1733 | NP_114123.2 | |
| XM_005260091.3 | 6 | 1900 | XP_005260148.1 | |
| XM_011528347.2 | 7 | 1762 | XP_011526649.1 | |
| XM_011528348.2 | 7 | 1936 | XP_011526650.1 | |
| XM_011528349.2 | 8 | 2395 | XP_011526651.1 | |
| XM_011528350.2 | 7 | 2247 | XP_011526652.1 | |
| XM_017027347.1 | 7 | 1920 | XP_016882836.1 | |
| AB054064.1 | 6 | 1733 | BAB91248.1 | |
| AF230330.1 | 6 | 1782 | AAK06404.1 | |
| AY358276.1 | 6 | 1658 | AAQ88643.1 | |
| BC142632.1 | 6 | 1651 |
Target Gene Details
Entrez Gene ID: | 55337 |
Gene Name: | chromosome 19 open reading frame 66 |
Gene Aliases: |
RyDEN |
Location: |
Chr.19:10086119-10093252 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C19orf66 | NM_001308277.1 | 8 | 1236 | NP_001295206.1 |
| NM_018381.3 | 8 | 1344 | NP_060851.2 | |
| XM_011528121.1 | 10 | 1514 | XP_011526423.1 | |
| XM_017026934.1 | 8 | 1282 | XP_016882423.1 | |
| AK002148.1 | 8 | 1130 | BAA92109.1 | |
| AK096142.1 | 8 | 1359 | BAC04710.1 | |
| AK098062.1 | 1 | 872 | ||
| AL137694.1 | 7 | 1168 | ||
| BC035817.2 | 8 | 1028 | AAH35817.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2414 | Chr.19:10063769 - 10097402 on Build GRCh38 | Insertion |
|
| nsv458355 | Chr.19:10061840 - 10102478 on Build GRCh38 | Loss |
|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map