Assay Details
Target Gene Details
Entrez Gene ID: | 9219 |
Gene Name: | metastasis associated 1 family member 2 |
Gene Aliases: |
MTA1L1, PID |
Location: |
Chr.11:62593203-62601840 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTA2 | NM_004739.3 | 4 | 650 | NP_004730.2 |
| XM_017018561.1 | 4 | 502 | XP_016874050.1 | |
| AB016591.1 | 4 | 583 | BAA36707.1 | |
| AF295807.1 | 4 | 260 | AAG02241.1 | |
| AK301569.1 | 4 | 485 | ||
| AL561056.3 | 4 | 649 | ||
| BC023656.2 | 4 | 446 | AAH23656.1 | |
| BC053650.1 | 4 | 468 | AAH53650.1 | |
| CR749481.1 | 2 | 171 | CAH18309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3584599 | Chr.11:62495270 - 62630578 on Build GRCh38 | Gain |
|
| nsv825939 | Chr.11:62558611 - 62643105 on Build GRCh38 | Gain |
|
| nsv832182 | Chr.11:62528103 - 62695376 on Build GRCh38 | Gain+Loss |
|
| nsv555174 | Chr.11:62566624 - 62729047 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map