Assay Details
Target Gene Details
Entrez Gene ID: | 2180 |
Gene Name: | acyl-CoA synthetase long-chain family member 1 |
Gene Aliases: |
ACS1, FACL1, FACL2, LACS, LACS1, LACS2 |
Location: |
Chr.4:184755595-184826593 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL1 | NM_001286708.1 | 21 | 3468 | NP_001273637.1 |
| NM_001286710.1 | 21 | 3630 | NP_001273639.1 | |
| NM_001286711.1 | 20 | 3465 | NP_001273640.1 | |
| NM_001286712.1 | 22 | 3813 | NP_001273641.1 | |
| NM_001995.3 | 21 | 3567 | NP_001986.2 | |
| XM_005262828.1 | 21 | 3395 | XP_005262885.1 | |
| XM_005262829.1 | 21 | 3462 | XP_005262886.1 | |
| XM_005262831.1 | 21 | 3579 | XP_005262888.1 | |
| XM_011531742.1 | 21 | 3495 | XP_011530044.1 | |
| XM_017007887.1 | 21 | 3437 | XP_016863376.1 | |
| XM_017007888.1 | 21 | 3521 | XP_016863377.1 | |
| XM_017007889.1 | 21 | 3579 | XP_016863378.1 | |
| AK096117.1 | 9 | 2124 | BAC04704.1 | |
| BC026290.1 | 16 | 2861 | AAH26290.1 | |
| BM982557.1 | 1 | 309 | ||
| D10040.1 | 20 | 3349 | BAA00931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830168 | Chr.4:184626297 - 184801578 on Build GRCh38 | Loss |
|
| nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map