Assay Details
Target Gene Details
Entrez Gene ID: | 84283 |
Gene Name: | transmembrane protein 79 |
Gene Aliases: |
MATT |
Location: |
Chr.1:156282913-156292443 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM79 | NM_032323.2 | 4 | 1599 | NP_115699.1 |
| NR_026678.1 | 4 | 1624 | ||
| AK056816.1 | 3 | 1840 | BAB71290.1 | |
| AK122634.1 | 5 | 1345 | ||
| BC005094.1 | 4 | 1589 | AAH05094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509524 | Chr.1:156238274 - 156308437 on Build GRCh38 | Insertion |
|
| nsv547968 | Chr.1:156282346 - 156294857 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map