Assay Details
Target Gene Details
Entrez Gene ID: | 9468 |
Gene Name: | phosphate cytidylyltransferase 1, choline, beta |
Gene Aliases: |
CCTB, CTB |
Location: |
Chr.X:24558087-24672862 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCYT1B | NM_001163264.1 | 8 | 2814 | NP_001156736.1 |
| NM_001163265.1 | 9 | 2546 | NP_001156737.1 | |
| NM_004845.4 | 8 | 2871 | NP_004836.2 | |
| XM_017029977.1 | 9 | 2696 | XP_016885466.1 | |
| AK226121.1 | 8 | 2814 | ||
| BC045634.1 | 8 | 2629 | AAH45634.2 |
More Information
Additional Information:
For this assay, SNP(s) [rs147766421] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map