Assay Details
Target Gene Details
Entrez Gene ID: | 65095 |
Gene Name: | KRI1 homolog |
Gene Aliases: |
- |
Location: |
Chr.19:10553085-10566031 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRI1 | NM_023008.3 | 19 | 2179 | NP_075384.3 |
| XM_011528190.2 | 18 | 2110 | XP_011526492.1 | |
| AK023011.1 | 17 | 1957 | BAB14357.1 | |
| AK025907.1 | 8 | 1033 | BAB15278.1 | |
| AK093879.1 | 18 | 2291 | BAC04240.1 | |
| AK097181.1 | 10 | 1497 | ||
| AK300780.1 | 19 | 2163 | ||
| AL833189.1 | 17 | 2918 | ||
| BC002890.2 | 16 | 1844 | AAH02890.3 | |
| BC009876.2 | 10 | 1351 | AAH09876.2 | |
| BC112249.1 | AAI12250.1 | |||
| BC112251.1 | AAI12252.1 | |||
| BC143770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map