Assay Details
Target Gene Details
Entrez Gene ID: | 160762 |
Gene Name: | coiled-coil domain containing 63 |
Gene Aliases: |
ODA5 |
Location: |
Chr.12:110844489-110907535 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC63 | NM_001286243.1 | NP_001273172.1 | ||
| NM_001286244.1 | NP_001273173.1 | |||
| NM_152591.2 | NP_689804.1 | |||
| XM_006719262.1 | XP_006719325.1 | |||
| XM_006719263.2 | XP_006719326.1 | |||
| XM_011537999.1 | XP_011536301.1 | |||
| XM_011538001.2 | XP_011536303.1 | |||
| AK093162.1 | BAC04081.1 | |||
| AK302207.1 | ||||
| AK310294.1 | ||||
| BC044815.1 | ||||
| BC064580.1 | AAH64580.1 | |||
| BC073147.1 | AAH73147.1 | |||
| DB036197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509476 | Chr.12:110775614 - 110881742 on Build GRCh38 | Insertion |
|
| esv3630744 | Chr.12:110849055 - 110980384 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map