Assay Details
Target Gene Details
Entrez Gene ID: | 83648 |
Gene Name: | family with sequence similarity 167 member A |
Gene Aliases: |
C8orf13, D8S265 |
Location: |
Chr.8:11421464-11475904 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM167A | NM_053279.2 | 3 | 1480 | NP_444509.2 |
| XM_005272398.4 | 4 | 1833 | XP_005272455.1 | |
| XM_011543837.1 | 3 | 1561 | XP_011542139.1 | |
| XM_011543838.2 | 3 | 1487 | XP_011542140.1 | |
| XM_011543840.2 | 3 | 1377 | XP_011542142.1 | |
| AJ301564.1 | 3 | 1389 | CAC82740.1 | |
| AK290704.1 | 3 | 1561 | ||
| AL157475.1 | 2 | 695 | CAB75670.1 | |
| AL834122.1 | 3 | 1469 | CAD38843.1 | |
| BC048214.1 | 2 | 669 | AAH48214.1 | |
| BC104041.1 | 2 | 1302 | AAI04042.1 | |
| BC104042.1 | 2 | 1302 | AAI04043.1 | |
| BC104043.1 | 2 | 1302 | AAI04044.1 |
Target Gene Details
Entrez Gene ID: | 83656 |
Gene Name: | FAM167A antisense RNA 1 |
Gene Aliases: |
C8orf12 |
Location: |
Chr.8:11368402-11438657 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM167A-AS1 | NR_026814.1 | |||
| AJ301563.1 | CAC82739.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34097 | Chr.8:11319863 - 11632385 on Build GRCh38 | Loss |
|
| nsv509252 | Chr.8:11419836 - 11481229 on Build GRCh38 | Insertion |
|
| nsv522860 | Chr.8:11396370 - 11472855 on Build GRCh38 | Loss |
|
| nsv465465 | Chr.8:11387555 - 11449609 on Build GRCh38 | Loss |
|
| nsv470183 | Chr.8:11379466 - 11449609 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map