Assay Details
Target Gene Details
Entrez Gene ID: | 7158 |
Gene Name: | tumor protein p53 binding protein 1 |
Gene Aliases: |
53BP1, TDRD30, TP53, p202, p53BP1 |
Location: |
Chr.15:43403064-43510509 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TP53BP1 | NM_001141979.1 | 28 | 6092 | NP_001135451.1 |
| NM_001141980.1 | 28 | 6098 | NP_001135452.1 | |
| NM_005657.2 | 28 | 6118 | NP_005648.1 | |
| XM_011521985.1 | 27 | 5947 | XP_011520287.1 | |
| XM_017022533.1 | 29 | 6202 | XP_016878022.1 | |
| AB210025.1 | 28 | 6063 | ||
| AF078776.1 | 28 | 6143 | AAC62018.1 | |
| AK123418.1 | 7 | 2104 | ||
| BC112161.1 | AAI12162.1 | |||
| BX537418.1 | 28 | 6097 | CAD97660.1 | |
| U09477.1 | 15 | 3137 | AAA21596.1 |
Target Gene Details
Entrez Gene ID: | 27229 |
Gene Name: | tubulin gamma complex associated protein 4 |
Gene Aliases: |
76P, GCP-4, GCP4, Grip76, MCCRP3 |
Location: |
Chr.15:43371059-43409771 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TUBGCP4 | XM_011521454.2 | 17 | 4301 | XP_011519756.1 |
| XM_011521455.2 | 18 | 4316 | XP_011519757.1 | |
| XM_017022078.1 | 17 | 4298 | XP_016877567.1 |
More Information
Set Membership: |
|

Genomic Map