Assay Details
Target Gene Details
Entrez Gene ID: | 219983 |
Gene Name: | olfactory receptor family 4 subfamily D member 6 |
Gene Aliases: |
OR11-250 |
Location: |
Chr.11:59456961-59457905 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR4D6 | NM_001004708.1 | NP_001004708.1 | ||
| BC137028.1 | 1 | 84 | ||
| BC137029.1 | 1 | 84 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825934 | Chr.11:59457040 - 59457741 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map