Assay Details
Target Gene Details
Entrez Gene ID: | 80125 |
Gene Name: | coiled-coil domain containing 33 |
Gene Aliases: |
CT61, HP11097 |
Location: |
Chr.15:74202705-74336472 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC33 | XM_017022629.1 | XP_016878118.1 |
Target Gene Details
Entrez Gene ID: | 64220 |
Gene Name: | stimulated by retinoic acid 6 |
Gene Aliases: |
MCOPCB8, MCOPS9, PP14296 |
Location: |
Chr.15:74179466-74212267 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRA6 | NM_001142617.1 | NP_001136089.1 | ||
| NM_001142618.1 | NP_001136090.1 | |||
| NM_001142619.1 | NP_001136091.1 | |||
| NM_001142620.1 | NP_001136092.1 | |||
| NM_001199040.1 | NP_001185969.1 | |||
| NM_001199041.1 | NP_001185970.1 | |||
| NM_001199042.1 | 1 | 155 | NP_001185971.1 | |
| NM_022369.3 | NP_071764.3 | |||
| XM_011521883.1 | XP_011520185.1 | |||
| XM_011521884.1 | XP_011520186.1 | |||
| XM_017022479.1 | XP_016877968.1 | |||
| XM_017022480.1 | XP_016877969.1 | |||
| AF352729.1 | AAK30290.1 | |||
| AF370419.2 | AAQ15255.2 | |||
| AK056125.1 | ||||
| AK091152.1 | ||||
| AK291966.1 | ||||
| AK298928.1 | ||||
| AK299191.1 | ||||
| AK302932.1 | ||||
| AK309676.1 | 1 | 135 | ||
| AK310878.1 | ||||
| AY358748.1 | AAQ89108.1 | |||
| BC015881.2 | AAH15881.1 | |||
| BC025256.1 | AAH25256.1 | |||
| CB131492.1 | ||||
| DA700543.1 | ||||
| DA746408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv471253 | Chr.15:74196866 - 74252807 on Build GRCh38 | Loss |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map