Assay Details
Target Gene Details
Entrez Gene ID: | 29945 |
Gene Name: | anaphase promoting complex subunit 4 |
Gene Aliases: |
APC4 |
Location: |
Chr.4:25377226-25418498 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANAPC4 | NM_001286756.1 | NP_001273685.1 | ||
| NM_013367.2 | NP_037499.2 | |||
| XM_011513838.1 | XP_011512140.1 | |||
| XM_011513839.1 | XP_011512141.1 | |||
| XM_017008138.1 | XP_016863627.1 | |||
| AF191338.1 | AAF05752.1 | |||
| AK023597.1 | ||||
| AK292336.1 | ||||
| BC059383.1 | AAH59383.1 | |||
| BI546948.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997544 | Chr.4:25362237 - 25915432 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map