Assay Details
Target Gene Details
Entrez Gene ID: | 55057 |
Gene Name: | absent in melanoma 1-like |
Gene Aliases: |
CRYBG2 |
Location: |
Chr.1:26321859-26354130 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AIM1L | NM_001039775.3 | 2 | 1395 | NP_001034864.2 |
| XM_005245918.2 | 2 | 1381 | XP_005245975.1 | |
| XM_011541672.1 | 2 | 1381 | XP_011539974.1 | |
| XM_011541673.2 | 2 | 1620 | XP_011539975.1 | |
| AK000902.1 | 2 | 1395 | BAA91416.1 | |
| CD628848.1 | 1 | 318 | ||
| CD628851.1 | 1 | 305 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745685 | Chr.1:26344350 - 26345589 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map