Assay Details
Target Gene Details
Entrez Gene ID: | 23580 |
Gene Name: | CDC42 effector protein 4 |
Gene Aliases: |
BORG4, CEP4, KAIA1777 |
Location: |
Chr.17:73283624-73312004 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC42EP4 | NM_012121.4 | 2 | 1258 | NP_036253.2 |
| XM_005257182.2 | 2 | 1573 | XP_005257239.1 | |
| AB042237.1 | 2 | 1181 | BAB17272.1 | |
| AF099664.1 | 1 | 1112 | AAD16299.1 | |
| AK027385.1 | 2 | 1249 | ||
| AK097835.1 | 3 | 1037 | ||
| AK312536.1 | ||||
| BC002774.1 | 2 | 1258 | AAH02774.1 | |
| BC010451.1 | 2 | 1234 | AAH10451.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833534 | Chr.17:73155642 - 73349355 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map