Assay Details
Target Gene Details
Entrez Gene ID: | 3491 |
Gene Name: | cysteine rich angiogenic inducer 61 |
Gene Aliases: |
CCN1, GIG1, IGFBP10 |
Location: |
Chr.1:85580761-85583967 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYR61 | NM_001554.4 | 5 | 1506 | NP_001545.2 |
| AF003114.1 | 4 | 1154 | AAF21597.1 | |
| AF003594.1 | 5 | 1405 | AAB61240.1 | |
| AF031385.1 | 5 | 1362 | AAB84227.1 | |
| AK096420.1 | 1 | 1335 | ||
| AK223385.1 | BAD97105.1 | |||
| AL832891.1 | 1 | 1153 | ||
| BC001271.1 | 5 | 1441 | AAH01271.1 | |
| BC009199.2 | 5 | 1490 | AAH09199.1 | |
| BC016952.1 | 5 | 1497 | AAH16952.1 | |
| U62015.1 | 5 | 1491 | AAB58319.1 | |
| Y11307.1 | 5 | 1505 | CAA72167.1 | |
| Y12084.1 | 5 | 1496 | CAA72802.1 | |
| Z98053.1 | 5 | 1500 | CAB10848.1 |
More Information
Set Membership: |
|

Genomic Map