Assay Details
Target Gene Details
Entrez Gene ID: | 65059 |
Gene Name: | Ras association (RalGDS/AF-6) and pleckstrin homology domains 1 |
Gene Aliases: |
ALS2CR18, ALS2CR9, LPD, PREL-2, PREL2, RMO1, RalGDS/AF-6 |
Location: |
Chr.2:203433682-203535335 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAPH1 | NM_213589.2 | 14 | 7784 | NP_998754.1 |
| XM_005246777.3 | 16 | 7935 | XP_005246834.1 | |
| XM_005246779.4 | 16 | 8834 | XP_005246836.1 | |
| XM_006712695.3 | 17 | 8915 | XP_006712758.1 | |
| XM_011511646.2 | 17 | 8007 | XP_011509948.1 | |
| XM_011511647.1 | 14 | 7640 | XP_011509949.1 | |
| XM_011511648.1 | 14 | 7634 | XP_011509950.1 | |
| AB053320.1 | 1 | 1834 | ||
| AJ584699.1 | 14 | 7778 | CAE48361.1 | |
| AK022729.1 | 1 | 2810 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001799 | Chr.2:202908732 - 205615330 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map