Assay Details
Target Gene Details
Entrez Gene ID: | 79170 |
Gene Name: | proline rich 15 like |
Gene Aliases: |
ATAD4 |
Location: |
Chr.17:47951967-47957905 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRR15L | NM_024320.3 | 2 | 1091 | NP_077296.1 |
| XM_005257663.4 | 2 | 1121 | XP_005257720.1 | |
| XM_005257664.2 | 1 | 877 | XP_005257721.1 | |
| XM_005257665.1 | 1 | 877 | XP_005257722.1 | |
| XM_005257666.4 | 3 | 1202 | XP_005257723.1 | |
| AA707319.1 | 1 | 330 | ||
| AK309589.1 | 3 | 642 | ||
| BC002865.2 | 2 | 958 | AAH02865.1 |

Genomic Map