Assay Details
Target Gene Details
Entrez Gene ID: | 2669 |
Gene Name: | GTP binding protein overexpressed in skeletal muscle |
Gene Aliases: |
KIR |
Location: |
Chr.8:94249249-94262345 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GEM | NM_005261.3 | 5 | 1662 | NP_005252.1 |
| NM_181702.2 | 5 | 1547 | NP_859053.1 | |
| XM_017013315.1 | 5 | 1866 | XP_016868804.1 | |
| BC022010.2 | 5 | 1547 | AAH22010.1 | |
| U10550.1 | 5 | 1636 | AAA64911.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7270n100 | Chr.8:94249727 - 94272186 on Build GRCh38 | Gain |
|
| nsv831401 | Chr.8:94206069 - 94381740 on Build GRCh38 | Loss |
|
| nsv1022473 | Chr.8:91854892 - 94250166 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map