Assay Details
Target Gene Details
Entrez Gene ID: | 51574 |
Gene Name: | La ribonucleoprotein domain family member 7 |
Gene Aliases: |
ALAZS, HDCMA18P, PIP7S |
Location: |
Chr.4:112636964-112657592 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP7 | NM_001267039.1 | 1 | 52 | NP_001253968.1 |
| NM_016648.3 | 1 | 52 | NP_057732.2 | |
| NR_049768.1 | 1 | 52 | ||
| DB270203.1 | 1 | 52 |
Target Gene Details
Entrez Gene ID: | 55345 |
Gene Name: | zinc finger GRF-type containing 1 |
Gene Aliases: |
C4orf21 |
Location: |
Chr.4:112539333-112636995 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZGRF1 | NM_018392.4 | NP_060862.3 | ||
| XM_011532091.2 | XP_011530393.1 | |||
| XM_011532093.2 | XP_011530395.1 | |||
| XM_011532094.2 | XP_011530396.1 | |||
| XM_011532096.2 | XP_011530398.1 | |||
| XM_017008370.1 | XP_016863859.1 | |||
| AK299831.1 | ||||
| AK302613.1 | ||||
| DB219969.1 | 1 | 448 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006030 | Chr.4:112618215 - 112736658 on Build GRCh38 | Gain |
|
| nsv525715 | Chr.4:112615590 - 112657453 on Build GRCh38 | Loss |
|
| nsv1014926 | Chr.4:112574087 - 112770975 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map