Assay Details
Target Gene Details
Entrez Gene ID: | 150468 |
Gene Name: | cytoskeleton associated protein 2 like |
Gene Aliases: |
- |
Location: |
Chr.2:112736349-112764677 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CKAP2L | NM_001304361.1 | 9 | 2399 | NP_001291290.1 |
| NM_152515.4 | 9 | 2458 | NP_689728.3 | |
| NR_130712.1 | 10 | 1700 | ||
| XM_011510666.2 | 8 | 2331 | XP_011508968.1 | |
| AK097948.1 | 10 | 1640 | BAC05202.1 | |
| AK292530.1 | 9 | 2409 | ||
| AK302875.1 | 9 | 2340 | ||
| AL832036.1 | 9 | 2409 | ||
| BC036217.1 | 9 | 2390 | AAH36217.1 |
Target Gene Details
Entrez Gene ID: | 284958 |
Gene Name: | 5'-nucleotidase domain containing 4 |
Gene Aliases: |
- |
Location: |
Chr.2:112717245-112742879 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NT5DC4 | XM_011512262.2 | XP_011510564.1 | ||
| XM_011512265.2 | XP_011510567.1 | |||
| XM_017005474.1 | XP_016860963.1 | |||
| XM_017005475.1 | XP_016860964.1 | |||
| XM_017005476.1 | XP_016860965.1 | |||
| XM_017005477.1 | XP_016860966.1 | |||
| XM_017005478.1 | XP_016860967.1 | |||
| BC041437.1 | AAH41437.1 |
More Information
Set Membership: |
|

Genomic Map