Assay Details
Target Gene Details
Entrez Gene ID: | 154091 |
Gene Name: | solute carrier family 2 member 12 |
Gene Aliases: |
GLUT12, GLUT8 |
Location: |
Chr.6:133987581-134052668 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC2A12 | NM_145176.2 | 5 | 2005 | NP_660159.1 |
| AK056554.1 | 4 | 744 | BAB71214.1 | |
| AK122628.1 | 5 | 1997 | ||
| AK226098.1 | 5 | 1876 | ||
| AL833602.1 | 5 | 1989 | ||
| AY046419.1 | 5 | 1967 | AAL02327.1 | |
| BC070149.1 | 5 | 1939 | AAH70149.1 | |
| BX648889.1 | 6 | 1140 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34122 | Chr.6:133767881 - 134057114 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map