Assay Details
Target Gene Details
Entrez Gene ID: | 11168 |
Gene Name: | PC4 and SFRS1 interacting protein 1 |
Gene Aliases: |
DFS70, LEDGF, PAIP, PSIP2, p52, p75 |
Location: |
Chr.9:15464066-15511019 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSIP1 | NM_001128217.2 | 16 | 2511 | NP_001121689.1 |
| NM_033222.4 | 16 | 2514 | NP_150091.2 | |
| AF063020.1 | 16 | 2480 | AAC25167.1 | |
| AF098483.1 | 15 | 2194 | AAC97946.1 | |
| AF432220.1 | 16 | 2462 | AAL99926.1 | |
| BX649155.1 | 6 | 3897 | ||
| U94319.1 | 10 | 1633 | AAB52589.1 |
Target Gene Details
Entrez Gene ID: | 6619 |
Gene Name: | small nuclear RNA activating complex polypeptide 3 |
Gene Aliases: |
PTFbeta, SNAP50 |
Location: |
Chr.9:15422696-15466751 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNAPC3 | XM_017015056.1 | XP_016870545.1 | ||
| XM_017015057.1 | XP_016870546.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517975 | Chr.9:15286448 - 15534072 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78732111] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map