Assay Details
Target Gene Details
Entrez Gene ID: | 340706 |
Gene Name: | von Willebrand factor A domain containing 2 |
Gene Aliases: |
AMACO, CCSP-2, NET42 |
Location: |
Chr.10:114239254-114294500 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWA2 | NM_001272046.1 | 4 | 507 | NP_001258975.1 |
| NM_001320804.1 | 4 | 449 | NP_001307733.1 | |
| XM_011539754.2 | 4 | 506 | XP_011538056.1 | |
| XM_011539757.2 | 4 | 779 | XP_011538059.1 | |
| XM_017016177.1 | 4 | 259 | XP_016871666.1 | |
| XM_017016178.1 | XP_016871667.1 | |||
| XM_017016179.1 | 4 | 212 | XP_016871668.1 | |
| XM_017016181.1 | 4 | 256 | XP_016871670.1 | |
| AJ536328.2 | 4 | 431 | CAD60276.1 | |
| AJ616914.1 | CAE83814.1 | |||
| AK122716.1 | 4 | 502 | BAC85505.1 | |
| AK127756.1 | 4 | 338 | BAC87116.1 | |
| AY572972.1 | 4 | 269 | AAT77225.1 | |
| BC128588.1 | ||||
| DA170480.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75028145] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map