Assay Details
Target Gene Details
Entrez Gene ID: | 57085 |
Gene Name: | angiotensin II receptor associated protein |
Gene Aliases: |
ATRAP |
Location: |
Chr.1:11736085-11750771 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGTRAP | NM_001040194.1 | 4 | 427 | NP_001035284.1 |
| NM_001040195.1 | 3 | 392 | NP_001035285.1 | |
| NM_001040196.1 | 5 | 524 | NP_001035286.1 | |
| NM_001040197.1 | 5 | 524 | NP_001035287.1 | |
| NM_020350.4 | 4 | 427 | NP_065083.3 | |
| XM_011541799.2 | 5 | 441 | XP_011540101.2 | |
| XM_011541800.2 | 5 | 441 | XP_011540102.1 | |
| XM_011541802.1 | 6 | 538 | XP_011540104.1 | |
| AF165187.1 | 4 | 303 | AAF89547.1 | |
| AF312374.1 | 4 | 358 | AAL26806.1 | |
| AK290042.1 | 4 | 370 | ||
| BC017328.2 | 4 | 365 | AAH17328.1 | |
| BI117521.1 | 4 | 399 | ||
| CA866314.1 | 3 | 398 | ||
| CD674964.1 | 5 | 404 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv545418 | Chr.1:11743229 - 11749469 on Build GRCh38 | Loss |
|
| nsv545419 | Chr.1:11747558 - 11751216 on Build GRCh38 | Gain |
|
| nsv545420 | Chr.1:11747913 - 11751226 on Build GRCh38 | Loss |
|
| nsv508880 | Chr.1:11721383 - 11771263 on Build GRCh38 | Insertion |
|
| nsv832758 | Chr.1:11637026 - 11777087 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map