Assay Details
Target Gene Details
Entrez Gene ID: | 392490 |
Gene Name: | TPT1-like protein |
Gene Aliases: |
- |
Location: |
Chr.X:72144184-72161750 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FLJ44635 | NM_207422.2 | 2 | 467 | NP_997305.2 |
| AK126598.1 | 2 | 467 | BAC86606.1 | |
| AY572224.1 | 2 | 467 | AAS76644.1 | |
| BC137552.1 | 2 | 223 | ||
| BC137564.1 | 2 | 223 |
More Information
Set Membership: |
|

Genomic Map