Assay Details
Target Gene Details
Entrez Gene ID: | 24139 |
Gene Name: | echinoderm microtubule associated protein like 2 |
Gene Aliases: |
ELP70, EMAP-2, EMAP2 |
Location: |
Chr.19:45609396-45645601 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EML2 | NM_001193268.1 | NP_001180197.1 | ||
| NM_001193269.1 | NP_001180198.1 | |||
| NM_012155.2 | NP_036287.1 | |||
| NR_034098.1 | ||||
| XM_006723125.2 | XP_006723188.1 | |||
| XM_006723126.2 | XP_006723189.1 | |||
| XM_011526694.2 | XP_011524996.1 | |||
| XM_011526695.2 | XP_011524997.1 | |||
| XM_011526698.2 | XP_011525000.1 | |||
| XM_011526700.2 | XP_011525002.1 | |||
| XM_011526701.2 | XP_011525003.1 | |||
| XM_017026550.1 | XP_016882039.1 | |||
| AB209773.1 | BAD93010.1 | |||
| AF103939.1 | AAD19904.1 | |||
| AK026616.1 | ||||
| AK225043.1 | ||||
| AK294668.1 | ||||
| AK295905.1 | ||||
| AK296258.1 | ||||
| AL096717.1 | CAB46373.2 | |||
| BC032630.1 | AAH32630.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833848 | Chr.19:45457372 - 45644939 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map