Assay Details
Target Gene Details
Entrez Gene ID: | 10992 |
Gene Name: | splicing factor 3b subunit 2 |
Gene Aliases: |
Cus1, SAP145, SF3B145, SF3b1, SF3b150 |
Location: |
Chr.11:66051772-66068911 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3B2 | NM_006842.2 | 19 | 2304 | NP_006833.2 |
| XM_005273726.3 | 19 | 2874 | XP_005273783.1 | |
| XM_011544740.2 | 19 | 2874 | XP_011543042.1 | |
| XM_017017144.1 | 19 | 2871 | XP_016872633.1 | |
| AK290850.1 | 19 | 2288 | ||
| AK300016.1 | 13 | 1578 | ||
| BC000401.2 | 19 | 2263 | AAH00401.2 | |
| BC007610.1 | 12 | 1487 | AAH07610.1 | |
| BC014125.2 | 7 | 692 | AAH14125.2 | |
| BC053577.1 | 19 | 2271 | AAH53577.1 | |
| BX537771.1 | 17 | 1977 | CAD97834.1 | |
| U41371.1 | 19 | 2243 | AAA97461.1 |
More Information
Set Membership: |
|

Genomic Map