Assay Details
Target Gene Details
Entrez Gene ID: | 27089 |
Gene Name: | ubiquinol-cytochrome c reductase complex III subunit VII |
Gene Aliases: |
MC3DN4, QCR8, QP-C, QPC, UQCR7 |
Location: |
Chr.5:132866627-132868844 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UQCRQ | NM_014402.4 | 3 | 1474 | NP_055217.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5004 | Chr.5:132862410 - 132869128 on Build GRCh38 | Insertion |
|
| nsv830489 | Chr.5:132815845 - 133000744 on Build GRCh38 | Loss |
|
| esv3606811 | Chr.5:132860904 - 132910818 on Build GRCh38 | Gain |
|
| nsv5003 | Chr.5:132845164 - 132889919 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs67367845] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map